Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018648.3 | NP_061118.1 | 2 | 251 | ||
GenBank mRNA | AB043104.1 | - | 2 | 261 | ||
BC008886.2 | - | 2 | 196 | 4308721 | ||
BC063023.1 | - | 2 | 222 | 6018963 | ||
BM554889.1 | - | 2 | 192 | |||
BQ218536.1 | - | 2 | 267 |