Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM55A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018710.2 | NP_061180.1 | 2 | 449 | ||
XM_011517152.2 | XP_011515454.1 | 2 | 449 | |||
GenBank mRNA | AK313783.1 | - | 2 | 362 | ||
AX775961.1 | - | 2 | 299 | |||
BC033892.1 | - | 2 | 299 | 4820065 | ||
CR749733.1 | - | 1 | 13 | |||
DA171968.1 | - | 2 | 449 |