Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 11 | 1690 | ||
GenBank mRNA | AJ505950.1 | - | 11 | 1558 | ||
AK023815.1 | - | 2 | 140 | |||
AK127135.1 | - | 11 | 1659 | |||
AL096739.1 | - | 4 | 834 | |||
BC007224.1 | - | 4 | 490 | |||
BC072450.1 | - | 7 | 990 | |||
BC098444.1 | - | 6 | 679 |