Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017564.9 | NP_060034.9 | 27, 28 | 3179 | ||
XM_011538537.2 | XP_011536839.1 | 27, 28 | 3220 | |||
XM_011538538.2 | XP_011536840.1 | 27, 28 | 3220 | |||
XM_011538539.2 | XP_011536841.1 | 13, 14 | 1423 | |||
XM_011538541.2 | XP_011536843.1 | 5, 6 | 530 | |||
XM_017019585.1 | XP_016875074.1 | 27, 28 | 3220 | |||
GenBank mRNA | AB052958.1 | - | 27, 28 | 3175 | ||
AJ295695.1 | - | 27, 28 | 3179 | |||
AK074051.1 | - | 5, 6 | 550 | |||
AK160380.1 | - | 26, 27 | 2892 | |||
AY311388.1 | - | 27, 28 | 3197 |