Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 5 | 731 | ||
GenBank mRNA | AK056388.1 | - | 1 | 31 | ||
AK293986.1 | - | 5 | 354 | |||
AY112698.1 | - | 5 | 706 | |||
AY948200.1 | - | 5 | 714 | |||
BC000848.1 | - | 1 | 65 | |||
BC061583.1 | - | 1 | 43 | |||
BC070158.1 | - | 4 | 256 | |||
BC087836.1 | - | 3 | 142 | |||
BC107057.2 | - | 6 | 399 | |||
BC148185.2 | - | 5 | 408 |