Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 8 | 984 | ||
GenBank mRNA | AB036706.1 | - | 8 | 896 | ||
AF271386.1 | - | 8 | 907 | |||
AK000029.1 | - | 8 | 933 | |||
AK222533.1 | - | 8 | 971 | |||
AY065972.1 | - | 8 | 974 | |||
AY157361.1 | - | 8 | 986 | |||
AY157362.1 | - | 8 | 978 | |||
AY358359.1 | - | 8 | 970 | |||
AY549722.1 | - | 8 | 1073 | |||
AY619691.1 | - | 8 | 894 | |||
AY619692.1 | - | 8 | 951 | |||
BC020664.1 | - | 8 | 984 | 4693329 | ||
CR457224.1 | - | 7 | 868 |