Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286721.1 | NP_001273650.1 | 5 | 943 | ||
NM_017664.3 | NP_060134.2 | 4 | 849 | |||
NR_104586.1 | 5 | 981 | ||||
NR_104587.1 | 5 | 943 | ||||
GenBank mRNA | AK000100.1 | - | 4 | 811 | ||
AK094222.1 | - | 1 | 849 | |||
BC001727.1 | - | 5 | 842 | 3354202 | ||
BC039715.1 | - | 5 | 911 | |||
BX354283.2 | - | 5 | 840 | |||
CB242465.1 | - | 1 | 364 |