Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FERMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017671.4 | NP_060141.3 | 8 | 1775 | ||
GenBank mRNA | AB105105.1 | - | 8 | 1775 | ||
AF443278.1 | - | 8 | 1344 | |||
AK025365.1 | - | 3 | 156 | |||
AK092195.1 | - | 6 | 833 | |||
AK314394.1 | - | 8 | 1165 | |||
AY137240.1 | - | 7 | 986 | |||
BC035882.1 | - | 8 | 1458 | 4941567 | ||
BC040545.1 | - | 7 | 1030 | 5277060 | ||
BX647465.1 | - | 3 | 182 |