Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143778.1 | NP_001137250.1 | 6 | 732 | ||
NM_017707.3 | NP_060177.2 | 7 | 759 | |||
XM_011541755.1 | XP_011540057.1 | 7 | 693 | |||
XM_017001685.1 | XP_016857174.1 | 7 | 693 | |||
XM_017001686.1 | XP_016857175.1 | 7 | 693 | |||
XM_017001687.1 | XP_016857176.1 | 7 | 666 | |||
XM_017001688.1 | XP_016857177.1 | 7 | 655 | |||
XM_017001689.1 | XP_016857178.1 | 7 | 710 | |||
GenBank mRNA | AB056749.1 | - | 7 | 680 | ||
AK092291.1 | - | 7 | 681 | |||
AK295100.1 | - | 6 | 654 | |||
AK299359.1 | - | 7 | 655 | |||
BC060786.1 | - | 7 | 707 | 5298355 |