Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013841.1 | NP_001013863.1 | 2 | 256 | ||
NM_017720.2 | NP_060190.2 | 2 | 256 | |||
XM_011528123.1 | XP_011526425.1 | 2 | 283 | |||
GenBank mRNA | AJ245719.1 | - | 2 | 217 | ||
AK000241.1 | - | 2 | 227 | |||
AK125774.1 | - | 4 | 766 | |||
BC000795.1 | - | 2 | 207 | 3451782 | ||
BF677969.1 | - | 2 | 256 |