Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136035.2 | NP_001129507.1 | 16 | 1935 | ||
NM_001142554.1 | NP_001136026.1 | 14 | 1969 | |||
NM_017722.3 | NP_060192.1 | 15 | 2056 | |||
XM_006722793.2 | XP_006722856.1 | 15 | 2178 | |||
XM_011528124.1 | XP_011526426.1 | 16 | 1854 | |||
XM_011528125.1 | XP_011526427.1 | 17 | 2387 | |||
XM_017026944.1 | XP_016882433.1 | 17 | 2378 | |||
XM_017026945.1 | XP_016882434.1 | 16 | 2291 | |||
XM_017026946.1 | XP_016882435.1 | 14 | 2091 | |||
XM_017026947.1 | XP_016882436.1 | 17 | 2364 | |||
GenBank mRNA | AF075055.1 | - | 4 | 337 | ||
AF196479.1 | - | 15 | 1806 | |||
AK000251.1 | - | 16 | 1894 | |||
AK292377.1 | - | 16 | 1935 | |||
AK295245.1 | - | 15 | 1925 | |||
AK304636.1 | - | 15 | 1965 | |||
BC002492.2 | - | 15 | 1886 | 3051390 | ||
BC018302.1 | - | 14 | 1964 | 3463518 | ||
BC040126.1 | - | 15 | 1925 | 5730044 | ||
BF798026.1 | - | 3 | 427 |