Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193509.1 | NP_001180438.1 | 18 | 2310 | ||
NM_017735.4 | NP_060205.3 | 18 | 2557 | |||
XM_005264416.1 | XP_005264473.1 | 17 | 2421 | |||
XM_011532958.1 | XP_011531260.1 | 17 | 2445 | |||
GenBank mRNA | AK000665.1 | - | 13 | 1549 | ||
AK027570.1 | - | 18 | 2466 | |||
AK299196.1 | - | 18 | 2299 | |||
BC001248.1 | - | 17 | 2090 | |||
BC063791.1 | - | 18 | 2515 | 6500951 |