Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308097.1 | NP_001295026.1 | 12 | 1630 | ||
NM_017769.4 | NP_060239.2 | 13 | 1671 | |||
XM_011536925.2 | XP_011535227.1 | 13 | 1637 | |||
XM_011536926.1 | XP_011535228.1 | 12 | 1554 | |||
GenBank mRNA | AB037754.1 | - | 13 | 1623 | ||
AK000340.1 | - | 13 | 1636 | |||
AK022867.1 | - | 3 | 446 | |||
AK225346.1 | - | 13 | 1636 | |||
AK296872.1 | - | 12 | 1595 | |||
BC000973.2 | - | 13 | 1617 | 3450488 | ||
BC040179.1 | - | 1 | 927 |