Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017780.3 | NP_060250.2 | 21 | 5325 | ||
XM_011517553.2 | XP_011515855.1 | 21 | 5346 | |||
XM_011517554.2 | XP_011515856.1 | 21 | 5324 | |||
XM_011517555.2 | XP_011515857.1 | 21 | 5346 | |||
XM_011517560.2 | XP_011515862.1 | 21 | 5346 | |||
XM_017013612.1 | XP_016869101.1 | 21 | 5400 | |||
XM_017013613.1 | XP_016869102.1 | 21 | 5346 | |||
GenBank mRNA | AB037837.1 | - | 17 | 2547 |