Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYBU SIRNA Assays ›
Gene Symbol:
SYBU
Gene Name:
syntabulin
Gene Aliases:
GOLSYN, OCSYN, SNPHL
Chromosome Location:
Chr.8: 109574176 - 109691791 on Build GRCh38
UniGene ID:
Hs.390738
Species:
Human
siRNA ID
HSS124720
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYBU
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099743.1
NP_001093213.1
8
2127
NM_001099744.1
NP_001093214.1
8
2130
NM_001099745.1
NP_001093215.1
8
1966
NM_001099746.1
NP_001093216.1
6
1680
NM_001099747.1
NP_001093217.1
8
1884
NM_001099748.1
NP_001093218.1
8
1887
NM_001099749.1
NP_001093219.1
6
1601
NM_001099750.1
NP_001093220.1
9
2008
NM_001099751.1
NP_001093221.1
8
1973
NM_001099752.1
NP_001093222.1
8
1976
NM_001099753.1
NP_001093223.1
7
1774
NM_001099754.1
NP_001093224.1
7
1777
NM_001099755.1
NP_001093225.1
6
1572
NM_001099756.1
NP_001093226.1
6
1848
NM_017786.5
NP_060256.3
9
2005
XM_005250999.1
XP_005251056.1
5
1276
XM_005251000.1
XP_005251057.1
5
1285
XM_011517154.2
XP_011515456.1
4
4035
XM_017013614.1
XP_016869103.1
7
1696
XM_017013615.1
XP_016869104.1
7
1670
GenBank mRNA
AB040905.1
-
6
1430
AB195676.1
-
9
1938
AB195677.1
-
8
1817
AB195678.1
-
6
1534
AB195679.1
-
6
2333
AK000373.1
-
6
1659
AK055246.1
-
8
1897
AK090559.1
-
6
1567
AK091374.1
-
3
1620
AK096558.1
-
7
1772
AK290469.1
-
8
1966
AK297182.1
-
5
1270
AL713645.1
-
9
1899
AL834406.1
-
3
1045
AY705385.1
-
7
1614
BC012082.1
-
6
1547
4661631
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of insulin secretion involved in cellular response to glucose stimulus
protein binding
microtubule binding
syntaxin-1 binding
kinesin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline