Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 9, 10 | 1186 | ||
GenBank mRNA | AF193054.1 | - | 6, 7 | 744 | ||
AF336851.1 | - | 3, 4 | 420 | |||
AK022979.1 | - | 9, 10 | 1056 | |||
AK025891.1 | - | 5, 6 | 551 | |||
AK300135.1 | - | 9, 10 | 1065 | |||
AL832131.1 | - | 9, 10 | 1102 | |||
AY044431.1 | - | 9, 10 | 1125 | |||
BC009304.1 | - | 9, 10 | 1044 | |||
BC011927.2 | - | 9, 10 | 1040 | |||
BC016060.1 | - | 9, 10 | 1040 | |||
BC126287.1 | - | 9, 10 | 1175 | 8992003 | ||
BC130275.1 | - | 9, 10 | 1175 | 40146306 | ||
BC144125.1 | - | 9, 10 | 1175 | 9052645 | ||
EU718675.1 | - | 9, 10 | 1093 |