Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPLOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017921.3 | NP_060391.2 | 12 | 1468 | ||
XM_011524979.1 | XP_011523281.1 | 12 | 1463 | |||
XM_011524980.1 | XP_011523282.1 | 12 | 1478 | |||
XM_011524981.1 | XP_011523283.1 | 12 | 1478 | |||
XM_011524982.2 | XP_011523284.1 | 12 | 1478 | |||
XM_017024830.1 | XP_016880319.1 | 8 | 864 | |||
GenBank mRNA | AB040932.1 | - | 12 | 1375 | ||
AK000664.1 | - | 12 | 1377 | |||
AK023046.1 | - | 10 | 1104 | |||
AK023272.1 | - | 9 | 1047 | |||
AK294473.1 | - | 11 | 1345 | |||
AL834300.1 | - | 9 | 980 | |||
BC025930.1 | - | 12 | 1409 | 5017718 |