Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017925.6 | NP_060395.5 | 12 | 1761 | ||
NR_073201.3 | 12 | 1761 | ||||
XM_006716804.3 | XP_006716867.1 | 12 | 1729 | |||
XM_006716805.3 | XP_006716868.1 | 12 | 1729 | |||
XM_006716807.3 | XP_006716870.1 | 3 | 254 | |||
XM_011517959.2 | XP_011516261.1 | 12 | 1729 | |||
XM_017014917.1 | XP_016870406.1 | 12 | 1729 | |||
XM_017014918.1 | XP_016870407.1 | 13 | 1929 | |||
XM_017014919.1 | XP_016870408.1 | 13 | 1896 | |||
XM_017014920.1 | XP_016870409.1 | 12 | 1753 | |||
GenBank mRNA | AK125842.1 | - | 6 | 623 | ||
AK127734.1 | - | 1 | 37 | |||
BC144484.1 | - | 8 | 993 | |||
BC144485.1 | - | 8 | 993 |