Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322026.1 | NP_001308955.1 | 3, 4 | 834 | ||
NM_001322027.1 | NP_001308956.1 | 3, 4 | 834 | |||
NM_001322028.1 | NP_001308957.1 | 3, 4 | 834 | |||
NM_001322029.1 | NP_001308958.1 | 3, 4 | 834 | |||
NM_001322030.1 | NP_001308959.1 | 3, 4 | 834 | |||
NM_001322032.1 | NP_001308961.1 | 3, 4 | 834 | |||
NM_017926.3 | NP_060396.2 | 3, 4 | 834 | |||
NM_017972.3 | NP_060442.2 | 2, 3 | 727 | |||
NR_110314.1 | 2, 3 | 727 | ||||
XM_006720191.2 | XP_006720254.1 | 3, 4 | 807 | |||
XM_017021427.1 | XP_016876916.1 | 4, 5 | 1034 | |||
XM_017021428.1 | XP_016876917.1 | 4, 5 | 1034 | |||
XM_017021429.1 | XP_016876918.1 | 4, 5 | 1034 | |||
XM_017021430.1 | XP_016876919.1 | 4, 5 | 1034 | |||
XM_017021431.1 | XP_016876920.1 | 4, 5 | 1034 | |||
XM_017021432.1 | XP_016876921.1 | 4, 5 | 1034 | |||
XM_017021433.1 | XP_016876922.1 | 3, 4 | 807 | |||
GenBank mRNA | AB032978.1 | - | 3, 4 | 785 | ||
AK000895.1 | - | 2, 3 | 192 | |||
AK023523.1 | - | 2, 3 | 1435 | |||
BC038835.1 | - | 2, 3 | 423 | |||
BC058032.1 | - | 3, 4 | 784 | 4703539 |