Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033540.2 | NP_284941.2 | 7 | 857 | ||
XM_005247596.3 | XP_005247653.2 | 7 | 815 | |||
XM_011512963.2 | XP_011511265.1 | 4 | 443 | |||
GenBank mRNA | AF329637.1 | - | 6 | 731 | ||
AK000700.1 | - | 7 | 814 | |||
AK314306.1 | - | 7 | 814 | |||
BC020661.1 | - | 7 | 823 | |||
BC040557.1 | - | 7 | 857 | 5270347 | ||
CR627379.1 | - | 4 | 507 | |||
U95822.1 | - | 3 | 320 |