Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284280.1 | NP_001271209.1 | 8 | 2179 | ||
NM_001284281.1 | NP_001271210.1 | 6 | 1501 | |||
XM_005267842.2 | XP_005267899.1 | 9 | 1468 | |||
GenBank mRNA | AB095930.1 | - | 8 | 1843 | ||
AK000714.1 | - | 9 | 1502 | |||
AK024297.1 | - | 5 | 1056 | |||
BC022778.1 | - | 8 | 2179 | |||
BC038932.1 | - | 6 | 1501 | 5579749 | ||
BC072409.1 | - | 9 | 1546 | 6142109 | ||
BX248247.1 | - | 5 | 675 |