Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193513.1 | NP_001180442.1 | 6, 7 | 457 | ||
NM_001193514.1 | NP_001180443.1 | 5, 6 | 337 | |||
NM_001193515.1 | NP_001180444.1 | 4, 5 | 250 | |||
NM_017964.3 | NP_060434.2 | 5, 6 | 337 | |||
XM_006712048.3 | XP_006712111.1 | 3, 4 | 168 | |||
XM_011532959.2 | XP_011531261.1 | 6, 7 | 434 | |||
XM_011532960.2 | XP_011531262.1 | 5, 6 | 593 | |||
XM_011532961.2 | XP_011531263.1 | 4, 5 | 271 | |||
XM_011532962.2 | XP_011531264.1 | 3, 4 | 422 | |||
XM_011532963.2 | XP_011531265.1 | 6, 7 | 434 | |||
XM_017004462.1 | XP_016859951.1 | 3, 4 | 184 | |||
XM_017004463.1 | XP_016859952.1 | 6, 7 | 434 | |||
XM_017004464.1 | XP_016859953.1 | 6, 7 | 434 | |||
XM_017004465.1 | XP_016859954.1 | 6, 7 | 434 | |||
XM_017004466.1 | XP_016859955.1 | 5, 6 | 329 | |||
GenBank mRNA | AF173387.1 | - | 289 | |||
AK055663.1 | - | 5, 6 | 314 | |||
AK096672.1 | - | 4, 5 | 271 | |||
AK304225.1 | - | 4, 5 | 227 | |||
AL832755.1 | - | 2, 3 | 94 | |||
BC066903.1 | - | 6, 7 | 434 | 4801303 | ||
EF560719.1 | - | 5, 6 | 315 | |||
EF560726.1 | - | 5, 6 | 315 |