Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 5, 6 | 942 | ||
XM_011545162.1 | XP_011543464.1 | 5, 6 | 502 | |||
XM_011545164.2 | XP_011543466.1 | 5, 6 | 1810 | |||
GenBank mRNA | AK056361.1 | - | 4, 5 | 449 | ||
AK096644.1 | - | 4, 5 | 792 | |||
AK098336.1 | - | 6, 7 | 911 | |||
AY320283.1 | - | 5, 6 | 982 | |||
BC010096.1 | - | 5, 6 | 850 | 3502170 | ||
BC052577.1 | - | 5, 6 | 841 | 4130626 |