Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FRMD4A SIRNA Assays ›
Gene Symbol:
FRMD4A
Gene Name:
FERM domain containing 4A
Gene Aliases:
CCAFCA, FRMD4, bA295P9.4
Chromosome Location:
Chr.10: 13643706 - 14330952 on Build GRCh38
UniGene ID:
Hs.330463
Species:
Human
siRNA ID
HSS124817
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FRMD4A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318336.1
NP_001305265.1
16, 17
1804
NM_001318337.1
NP_001305266.1
16, 17
2044
NM_001318338.1
NP_001305267.1
6, 7
838
NM_018027.4
NP_060497.3
17, 18
1744
XM_005252489.2
XP_005252546.1
16, 17
2044
XM_005252490.2
XP_005252547.1
17, 18
1840
XM_005252495.4
XP_005252552.1
3, 4
3003
XM_006717457.2
XP_006717520.1
18, 19
2125
XM_006717458.3
XP_006717521.1
18, 19
2125
XM_006717459.2
XP_006717522.1
17, 18
2089
XM_006717460.2
XP_006717523.1
17, 18
2080
XM_006717464.1
XP_006717527.1
3, 4
504
XM_011519539.1
XP_011517841.1
18, 19
1885
XM_011519540.1
XP_011517842.1
18, 19
2125
XM_011519541.1
XP_011517843.1
18, 19
2125
XM_011519543.2
XP_011517845.1
18, 19
1848
XM_017016393.1
XP_016871882.1
17, 18
2089
XM_017016394.1
XP_016871883.1
17, 18
1812
XM_017016395.1
XP_016871884.1
16, 17
1647
XM_017016396.1
XP_016871885.1
5, 6
780
GenBank mRNA
AB037715.1
-
17, 18
1744
AK094045.1
-
16, 17
2045
AK289693.1
-
17, 18
1669
BC151244.1
-
17, 18
1755
8860518
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
establishment of epithelial cell polarity
protein binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-tl62x:80/100.66.78.111:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline