Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 14 | 1904 | ||
XM_005256038.3 | XP_005256095.1 | 14 | 1915 | |||
XM_005256041.3 | XP_005256098.2 | 14 | 1915 | |||
XM_011523223.2 | XP_011521525.1 | 14 | 1915 | |||
XM_011523224.2 | XP_011521526.1 | 14 | 1915 | |||
XM_011523225.2 | XP_011521527.1 | 14 | 1915 | |||
XM_017023441.1 | XP_016878930.1 | 14 | 1915 | |||
XM_017023442.1 | XP_016878931.1 | 14 | 1915 | |||
GenBank mRNA | AK001167.1 | - | 4 | 363 | ||
AK025479.1 | - | 8 | 937 | |||
AK056433.1 | - | 14 | 1914 | |||
AK297564.1 | - | 14 | 1689 | |||
AK307766.1 | - | 13 | 1754 | |||
AK310466.1 | - | 2 | 334 | |||
BC000536.2 | - | 4 | 368 | |||
BC007214.1 | - | 9 | 1026 | |||
BC110852.1 | - | 9 | 959 | |||
BC125108.1 | - | 14 | 1836 | 40118772 | ||
BC125109.1 | - | 14 | 1836 | 40118777 | ||
CR457343.1 | - | 8 | 872 |