Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 2 | 735 | ||
NM_001318717.1 | NP_001305646.1 | 2 | 656 | |||
NM_001318718.1 | NP_001305647.1 | 2 | 636 | |||
NM_001318719.1 | NP_001305648.1 | 2 | 614 | |||
NM_001318720.1 | NP_001305649.1 | 2 | 608 | |||
NM_001318721.1 | NP_001305650.1 | 2 | 583 | |||
NM_001318722.1 | NP_001305651.1 | 2 | 604 | |||
NM_001318723.1 | NP_001305652.1 | 2 | 688 | |||
NM_001318724.1 | NP_001305653.1 | 2 | 688 | |||
NM_001318725.1 | NP_001305654.1 | 2 | 591 | |||
NM_016506.6 | NP_057590.3 | 2 | 692 | |||
NM_018095.5 | NP_060565.4 | 2 | 587 | |||
XM_017018009.1 | XP_016873498.1 | 2 | 571 | |||
GenBank mRNA | AK001312.1 | - | 2 | 541 | ||
AK001749.1 | - | 2 | 628 | |||
AK091607.1 | - | 1 | 1265 | |||
AK222550.1 | - | 2 | 538 | |||
AK222985.1 | - | 2 | 548 | |||
AK225062.1 | - | 2 | 538 | |||
AL713735.1 | - | 1 | 517 | |||
BC002736.2 | - | 2 | 629 | 3628290 | ||
BI601113.1 | - | 2 | 594 | |||
BI908078.1 | - | 2 | 594 | |||
CR457270.1 | - | 1 | 475 |