Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018117.11 | NP_060587.8 | 24 | 3217 | ||
XM_005269962.2 | XP_005270019.1 | 24 | 2934 | |||
XM_005269963.2 | XP_005270020.1 | 24 | 3066 | |||
XM_005269964.2 | XP_005270021.1 | 17 | 1972 | |||
XM_017016397.1 | XP_016871886.1 | 23 | 2905 | |||
XM_017016398.1 | XP_016871887.1 | 23 | 2835 | |||
XM_017016399.1 | XP_016871888.1 | 23 | 2779 | |||
GenBank mRNA | AB037772.2 | - | 24 | 3030 | ||
AF320223.1 | - | 24 | 3004 | |||
AK000579.1 | - | 10 | 1019 | |||
AK001368.1 | - | 19 | 2120 | |||
AK025050.1 | - | 7 | 728 | |||
AK124522.1 | - | 11 | 3404 | |||
AK301761.1 | - | 4 | 417 | |||
AK315145.1 | - | 24 | 2995 | |||
AL137699.1 | - | 22 | 2690 | |||
BC040469.1 | - | 24 | 3034 | 4730308 | ||
BC071564.1 | - | 24 | 3000 | 30346203 |