Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160042.1 | NP_001153514.1 | 5 | 1465 | ||
NM_018134.2 | NP_060604.2 | 5 | 1225 | |||
GenBank mRNA | AK001409.1 | - | 5 | 1189 | ||
AK223272.1 | - | 5 | 1213 | |||
AK299235.1 | - | 4 | 1400 | |||
AK307853.1 | - | 5 | 1429 | |||
BC029986.1 | - | 1 | 353 | |||
BC098260.1 | - | 5 | 1188 | 40024457 | ||
BC098317.1 | - | 5 | 1188 | 40024460 | ||
BC098366.1 | - | 5 | 1188 | 40024462 |