Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018140.3 | NP_060610.2 | 9, 10 | 1618 | ||
XM_005248322.3 | XP_005248379.1 | 9, 10 | 1540 | |||
XM_011514063.1 | XP_011512365.1 | 9, 10 | 1564 | |||
XM_011514064.2 | XP_011512366.1 | 9, 10 | 1671 | |||
XM_017009626.1 | XP_016865115.1 | 8, 9 | 1452 | |||
XM_017009627.1 | XP_016865116.1 | 8, 9 | 1412 | |||
GenBank mRNA | AB040952.1 | - | 9, 10 | 1556 | ||
AK001427.1 | - | 9, 10 | 1561 | |||
BC000132.2 | - | 9, 10 | 1557 | 2899944 | ||
BC001750.1 | - | 9, 10 | 1544 |