Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 13, 14 | 1514 | ||
NM_001320048.1 | NP_001306977.1 | 11, 12 | 1269 | |||
NM_018186.3 | NP_060656.2 | 12, 13 | 1817 | |||
XM_005245317.4 | XP_005245374.1 | 12, 13 | 1389 | |||
XM_011509735.2 | XP_011508037.1 | 9, 10 | 999 | |||
XM_017001722.1 | XP_016857211.1 | 12, 13 | 1389 | |||
XM_017001723.1 | XP_016857212.1 | 12, 13 | 1389 | |||
XM_017001724.1 | XP_016857213.1 | 11, 12 | 1261 | |||
XM_017001725.1 | XP_016857214.1 | 10, 11 | 1122 | |||
GenBank mRNA | AK001568.1 | - | 11, 12 | 1189 | ||
AK023532.1 | - | 2, 3 | 252 | |||
AK096493.1 | - | 13, 14 | 1424 | |||
AK294569.1 | - | 8, 9 | 960 | |||
AK310638.1 | - | 8, 9 | 930 | |||
AL354614.1 | - | 12, 13 | 1817 | |||
BC091516.1 | - | 13, 14 | 1428 | 30389801 | ||
BC107168.1 | - | 11, 12 | 1166 | 40035373 | ||
BC107169.1 | - | 12, 13 | 1305 | 40035375 |