Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 17 | 1867 | ||
NM_001320048.1 | NP_001306977.1 | 15 | 1622 | |||
NM_018186.3 | NP_060656.2 | 16 | 2170 | |||
XM_005245317.4 | XP_005245374.1 | 16 | 1742 | |||
XM_011509735.2 | XP_011508037.1 | 13 | 1352 | |||
XM_017001722.1 | XP_016857211.1 | 16 | 1742 | |||
XM_017001723.1 | XP_016857212.1 | 16 | 1742 | |||
XM_017001724.1 | XP_016857213.1 | 15 | 1614 | |||
XM_017001725.1 | XP_016857214.1 | 14 | 1475 | |||
GenBank mRNA | AK001568.1 | - | 15 | 1542 | ||
AK023532.1 | - | 6 | 605 | |||
AK096493.1 | - | 17 | 1777 | |||
AK127098.1 | - | 20 | 2564 | |||
AK294569.1 | - | 12 | 1313 | |||
AK310638.1 | - | 12 | 1283 | |||
AL354614.1 | - | 16 | 2170 | |||
BC091516.1 | - | 17 | 1781 | 30389801 | ||
BC107168.1 | - | 15 | 1519 | 40035373 | ||
BC107169.1 | - | 16 | 1658 | 40035375 |