Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145025.1 | NP_001138497.1 | 9, 10 | 1217 | ||
NM_018217.2 | NP_060687.2 | 10, 11 | 1328 | |||
NR_026728.1 | 9, 10 | 1544 | ||||
GenBank mRNA | AK001645.1 | - | 10, 11 | 1310 | ||
AK023931.1 | - | 10, 11 | 1298 | |||
AK096016.1 | - | 8, 9 | 1238 | |||
AK293583.1 | - | 9, 10 | 1199 | |||
AK298794.1 | - | 9, 10 | 1286 | |||
AK300212.1 | - | 9, 10 | 1172 | |||
AK303816.1 | - | 8, 9 | 970 | |||
AY007088.1 | - | 7, 8 | 896 | |||
AY358580.1 | - | 9, 10 | 1178 | |||
BC001371.2 | - | 10, 11 | 1295 | 3051376 | ||
BC016184.1 | - | 10, 11 | 1269 | 4860424 | ||
CR457266.1 | - | 10, 11 | 1228 |