Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 18, 19 | 2495 | ||
NM_001143997.1 | NP_001137469.1 | 17, 18 | 2290 | |||
NM_018245.2 | NP_060715.2 | 19, 20 | 2666 | |||
XM_011539946.1 | XP_011538248.1 | 19, 20 | 2551 | |||
XM_011539947.1 | XP_011538249.1 | 19, 20 | 2757 | |||
XM_017016401.1 | XP_016871890.1 | 19, 20 | 2941 | |||
XM_017016402.1 | XP_016871891.1 | 19, 20 | 2568 | |||
XM_017016403.1 | XP_016871892.1 | 17, 18 | 2314 | |||
GenBank mRNA | AB033116.3 | - | 18, 19 | 2529 | ||
AK001713.1 | - | 19, 20 | 2610 | |||
AK055817.1 | - | 1, 2 | 1560 | |||
AK290226.1 | - | 18, 19 | 2639 | |||
AK296551.1 | - | 18, 19 | 2404 | |||
AK303729.1 | - | 17, 18 | 2237 | |||
BC026320.1 | - | 19, 20 | 2585 | 4797814 |