Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 22 | 2987 | ||
NM_001143997.1 | NP_001137469.1 | 21 | 2782 | |||
NM_018245.2 | NP_060715.2 | 23 | 3158 | |||
XM_011539946.1 | XP_011538248.1 | 23 | 3043 | |||
XM_011539947.1 | XP_011538249.1 | 23 | 3249 | |||
XM_017016401.1 | XP_016871890.1 | 23 | 3433 | |||
XM_017016402.1 | XP_016871891.1 | 23 | 3060 | |||
XM_017016403.1 | XP_016871892.1 | 21 | 2806 | |||
GenBank mRNA | AB033116.3 | - | 22 | 3021 | ||
AK001713.1 | - | 23 | 3102 | |||
AK055817.1 | - | 5 | 2052 | |||
AK290226.1 | - | 22 | 3131 | |||
AK296551.1 | - | 22 | 2896 | |||
AK303729.1 | - | 21 | 2729 | |||
BC026320.1 | - | 23 | 3077 | 4797814 |