Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018245.2 | NP_060715.2 | 3 | 466 | ||
XM_011539946.1 | XP_011538248.1 | 3 | 351 | |||
XM_011539947.1 | XP_011538249.1 | 3 | 557 | |||
XM_017016401.1 | XP_016871890.1 | 3 | 430 | |||
XM_017016402.1 | XP_016871891.1 | 3 | 341 | |||
XM_017016403.1 | XP_016871892.1 | 2 | 205 | |||
GenBank mRNA | AB033116.3 | - | 2 | 329 | ||
AK001713.1 | - | 3 | 410 | |||
AK225558.1 | - | 3 | 397 | |||
AK290226.1 | - | 2 | 439 | |||
AK307759.1 | - | 2 | 225 | |||
BC026320.1 | - | 3 | 385 | 4797814 | ||
DA238469.1 | - | 3 | 466 |