Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011649.2 | NP_001011649.1 | 24 | 3673 | ||
NM_001272039.1 | NP_001258968.1 | 21 | 2983 | |||
NM_018249.5 | NP_060719.4 | 24 | 3673 | |||
NR_073554.1 | 24 | 3750 | ||||
NR_073555.1 | 24 | 3673 | ||||
NR_073556.1 | 25 | 3880 | ||||
NR_073557.1 | 24 | 3753 | ||||
NR_073558.1 | 24 | 3750 | ||||
XM_006717182.1 | XP_006717245.1 | 23 | 3566 | |||
XM_006717185.1 | XP_006717248.1 | 21 | 2975 | |||
XM_011518860.1 | XP_011517162.1 | 24 | 3659 | |||
XM_017014921.1 | XP_016870410.1 | 23 | 3563 | |||
XM_017014922.1 | XP_016870411.1 | 18 | 2891 | |||
XM_017014923.1 | XP_016870412.1 | 21 | 2975 | |||
XM_017014924.1 | XP_016870413.1 | 15 | 2647 | |||
GenBank mRNA | AB046853.2 | - | 24 | 3642 | ||
AF448860.1 | - | 24 | 3623 | |||
AK001729.1 | - | 2 | 511 | |||
AK027636.1 | - | 3 | 637 | |||
AK122913.1 | - | 24 | 3662 | |||
AL133161.1 | - | 8 | 1663 | |||
AL833481.1 | - | 24 | 3661 | |||
BC136275.1 | - | 24 | 3505 | 9020262 | ||
BC143762.1 | - | 24 | 3508 | 9052276 | ||
BC146782.1 | - | 24 | 3642 | 9007237 | ||
BX537421.1 | - | 24 | 3658 | |||
BX537708.1 | - | 21 | 2972 | |||
BX537759.1 | - | 24 | 3661 | |||
BX640896.1 | - | 17 | 2789 | |||
CR936622.1 | - | 17 | 2803 | |||
CR936717.1 | - | 17 | 2803 | |||
KP294527.1 | - | 24 | 3673 |