Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020121.3 | NP_064506.3 | 29 | 3602 | ||
XM_011521094.1 | XP_011519396.1 | 30 | 3638 | |||
XM_011521095.1 | XP_011519397.1 | 29 | 3557 | |||
XM_011521096.1 | XP_011519398.1 | 29 | 3506 | |||
XM_011521097.1 | XP_011519399.1 | 30 | 3638 | |||
XM_011521098.1 | XP_011519400.1 | 23 | 2655 | |||
XM_011521099.2 | XP_011519401.1 | 22 | 2572 | |||
XM_011521102.2 | XP_011519404.1 | 20 | 2346 | |||
XM_011521103.2 | XP_011519405.1 | 20 | 2404 | |||
XM_017020662.1 | XP_016876151.1 | 28 | 3425 | |||
XM_017020663.1 | XP_016876152.1 | 30 | 3540 | |||
GenBank mRNA | AF227906.2 | - | 29 | 3503 | ||
AK001735.1 | - | 29 | 3527 | |||
AK021547.1 | - | 6 | 1075 | |||
AL133051.1 | - | 26 | 2982 | |||
BC125233.1 | - | 29 | 3458 | 40124676 |