Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
siRNA ID
HSS124988
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
23
3082
NM_001280545.1
NP_001267474.1
24
3254
NM_001280546.1
NP_001267475.1
21
2847
NM_018262.3
NP_060732.2
22
2926
NM_052985.3
NP_443711.2
24
3256
NM_052989.2
NP_443715.1
23
3103
NM_052990.2
NP_443716.1
20
2773
XM_005247609.1
XP_005247666.1
22
2894
XM_006713689.1
XP_006713752.1
23
3071
XM_006713691.2
XP_006713754.1
22
3017
XM_006713692.2
XP_006713755.1
21
2840
XM_017006817.1
XP_016862306.1
23
3071
XM_017006818.1
XP_016862307.1
23
3068
XM_017006819.1
XP_016862308.1
22
3017
XM_017006820.1
XP_016862309.1
22
3014
XM_017006821.1
XP_016862310.1
22
3014
XM_017006822.1
XP_016862311.1
21
2862
XM_017006823.1
XP_016862312.1
22
2894
XM_017006824.1
XP_016862313.1
22
2891
XM_017006825.1
XP_016862314.1
21
2840
XM_017006826.1
XP_016862315.1
21
2837
XM_017006827.1
XP_016862316.1
21
2837
XM_017006828.1
XP_016862317.1
20
2685
XM_017006829.1
XP_016862318.1
20
2682
XM_017006830.1
XP_016862319.1
24
3069
XM_017006831.1
XP_016862320.1
22
2990
XM_017006832.1
XP_016862321.1
23
2969
XM_017006833.1
XP_016862322.1
19
2631
XM_017006834.1
XP_016862323.1
19
2628
XM_017006835.1
XP_016862324.1
21
2813
XM_017006836.1
XP_016862325.1
22
2838
XM_017006837.1
XP_016862326.1
14
2015
XM_017006838.1
XP_016862327.1
19
2452
GenBank mRNA
AF244930.1
-
23
3000
AF244931.1
-
24
3153
AF302154.1
-
23
2885
AK001759.1
-
22
2739
AK001955.1
-
12
1551
AK022525.1
-
22
2837
AK023000.1
-
22
2568
AK024435.1
-
21
2614
AK094922.1
-
7
972
AK096891.1
-
8
1217
AK122965.1
-
16
2607
AK124140.1
-
24
3057
AK223095.1
-
22
2738
AK293852.1
-
19
2533
AK298526.1
-
19
2350
AK299903.1
-
21
2560
AK315103.1
-
23
2906
AL133565.1
-
21
2680
AL833851.1
-
12
1553
BC003045.1
-
20
2565
2823536
BC004238.2
-
20
2565
2823536
BC028353.2
-
23
2843
4836844
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline