Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018304.3 | NP_060774.2 | 2, 3 | 437 | ||
GenBank mRNA | AK001891.1 | - | 2, 3 | 190 | ||
AK225377.1 | - | 2, 3 | 190 | |||
AK225380.1 | - | 2, 3 | 195 | |||
AL711011.1 | - | 2, 3 | 204 | |||
BC008669.1 | - | 2, 3 | 179 | 3867293 | ||
DA649803.1 | - | 2, 3 | 437 | |||
DB106349.1 | - | 2, 3 | 204 | |||
FJ161961.1 | - | 2, 3 | 190 |