Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018368.3 | NP_060838.3 | 13 | 1585 | ||
XM_011535941.1 | XP_011534243.1 | 13 | 1435 | |||
GenBank mRNA | AF113224.1 | - | 13 | 1305 | ||
AF208863.1 | - | 12 | 1433 | |||
AF211480.1 | - | 10 | 923 | |||
AK002102.1 | - | 13 | 1334 | |||
AK225187.1 | - | 13 | 1334 | |||
AK225537.1 | - | 13 | 1319 | |||
AK290069.1 | - | 13 | 1465 | |||
AY136817.1 | - | 12 | 1090 | |||
BC010360.1 | - | 11 | 1084 | |||
BC047073.1 | - | 13 | 1424 | 5274221 | ||
BX647737.1 | - | 12 | 1177 |