Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018416.2 | NP_060886.1 | 3, 4 | 1489 | ||
XM_011520760.2 | XP_011519062.1 | 3, 4 | 1572 | |||
XM_011520761.2 | XP_011519063.1 | 4, 5 | 1673 | |||
GenBank mRNA | AF155132.1 | - | 3, 4 | 893 | ||
AF155133.1 | - | 3, 4 | 893 | |||
BC126396.1 | - | 3, 4 | 459 | 8992112 | ||
BC136305.1 | - | 3, 4 | 459 | 9020292 |