Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280801.1 | NP_001267730.1 | 6 | 1325 | ||
NM_018434.5 | NP_060904.2 | 6 | 1325 | |||
XM_011534593.2 | XP_011532895.1 | 6 | 1337 | |||
GenBank mRNA | AB209800.1 | - | 6 | 1308 | ||
AF155650.1 | - | 5 | 624 | |||
AK313089.1 | - | 6 | 808 | |||
AY083998.1 | - | 6 | 1307 | |||
BC017100.1 | - | 6 | 907 | |||
BC065244.1 | - | 5 | 570 | |||
BC082267.1 | - | 6 | 922 | |||
BC108306.1 | - | 6 | 1320 | 6137501 | ||
BC113864.1 | - | 7 | 755 | 40033418 | ||
BC114472.1 | - | 6 | 687 | |||
BP390145.1 | - | 1 | 41 | |||
BQ447530.1 | - | 4 | 546 |