Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280801.1 | NP_001267730.1 | 2 | 805 | ||
NM_018434.5 | NP_060904.2 | 2 | 805 | |||
XM_011534593.2 | XP_011532895.1 | 2 | 817 | |||
GenBank mRNA | AB209800.1 | - | 2 | 788 | ||
AK313089.1 | - | 2 | 288 | |||
AY083998.1 | - | 2 | 787 | |||
BC017100.1 | - | 2 | 387 | |||
BC065244.1 | - | 1 | 50 | |||
BC082267.1 | - | 2 | 457 | |||
BC108306.1 | - | 2 | 800 | 6137501 | ||
BC113864.1 | - | 3 | 235 | 40033418 | ||
BC114472.1 | - | 2 | 167 |