Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290185.1 | NP_001277114.1 | 5 | 1110 | ||
NM_021729.5 | NP_068375.3 | 5 | 895 | |||
XM_011542899.2 | XP_011541201.1 | 5 | 853 | |||
GenBank mRNA | AB027508.2 | - | 5 | 865 | ||
AF258558.1 | - | 3 | 396 | |||
AF308800.1 | - | 5 | 829 | |||
AK023279.1 | - | 5 | 866 | |||
AK056701.1 | - | 4 | 722 | |||
AK223158.1 | - | 5 | 855 | |||
AK302973.1 | - | 5 | 1042 | |||
AL834218.1 | - | 5 | 842 | |||
BC065563.1 | - | 5 | 829 | 6144541 |