Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018467.3 | NP_060937.1 | 3 | 229 | ||
XM_017026975.1 | XP_016882464.1 | 4 | 939 | |||
XM_017026976.1 | XP_016882465.1 | 2 | 179 | |||
GenBank mRNA | AB097050.1 | - | 3 | 200 | ||
AF220052.1 | - | 3 | 200 | |||
AK074683.1 | - | 3 | 185 | |||
BC006005.1 | - | 3 | 185 | 4279653 | ||
BC008455.1 | - | 3 | 177 | 4247241 | ||
BI758045.1 | - | 3 | 216 | |||
DA506853.1 | - | 2 | 179 |