Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163022.1 | NP_001156494.1 | 4 | 446 | ||
NM_001163023.1 | NP_001156495.1 | 3 | 356 | |||
NM_001276389.1 | NP_001263318.1 | 3 | 383 | |||
NM_018474.4 | NP_060944.3 | 5 | 609 | |||
XM_011529296.2 | XP_011527598.1 | 5 | 612 | |||
XM_011529297.2 | XP_011527599.1 | 5 | 601 | |||
XM_011529298.2 | XP_011527600.1 | 5 | 594 | |||
XM_011529299.2 | XP_011527601.1 | 4 | 441 | |||
XM_017027951.1 | XP_016883440.1 | 6 | 555 | |||
XM_017027952.1 | XP_016883441.1 | 4 | 441 | |||
XM_017027953.1 | XP_016883442.1 | 6 | 627 | |||
XM_017027954.1 | XP_016883443.1 | 4 | 441 | |||
GenBank mRNA | AF451990.1 | - | 5 | 587 | ||
AK025476.1 | - | 5 | 540 | |||
AK293166.1 | - | 3 | 287 | |||
AK293567.1 | - | 3 | 327 | |||
AK295355.1 | - | 4 | 345 | |||
BC039296.1 | - | 5 | 571 | |||
BC045826.1 | - | 4 | 333 | 5262165 | ||
BC065550.1 | - | 3 | 190 | |||
BC090879.2 | - | 1 | 14 | |||
BC105093.1 | - | 5 | 512 | 8144096 | ||
BC113370.1 | - | 5 | 512 | 8322422 | ||
DC386700.1 | - | 4 | 446 |