Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173487.1 | NP_001166958.1 | 4 | 1313 | ||
NM_001173488.1 | NP_001166959.1 | 2 | 1699 | |||
NM_017544.3 | NP_060014.2 | 3 | 1215 | |||
XM_011531365.2 | XP_011529667.1 | 4 | 1845 | |||
GenBank mRNA | AJ011812.2 | - | 2 | 1699 | ||
AY208891.1 | - | 3 | 1215 | |||
BC040379.1 | - | 3 | 1197 | 4777539 | ||
BC047878.2 | - | 3 | 1182 | 5228666 | ||
BC068514.1 | - | 3 | 1193 | 5299667 | ||
EF165092.1 | - | 1 | 773 | |||
EF165095.1 | - | 1 | 158 | |||
EF216688.1 | - | 2 | 1699 | |||
EF216689.1 | - | 2 | 1699 |