Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 4 | 363 | ||
NM_001034024.1 | NP_001029196.1 | 4 | 323 | |||
NM_019100.4 | NP_061973.1 | 3 | 544 | |||
XM_005271037.4 | XP_005271094.1 | 3 | 534 | |||
XM_005271039.1 | XP_005271096.1 | 4 | 434 | |||
XM_005271040.1 | XP_005271097.1 | 4 | 311 | |||
XM_006710771.3 | XP_006710834.1 | 2 | 213 | |||
XM_011541784.2 | XP_011540086.1 | 4 | 368 | |||
XM_011541785.2 | XP_011540087.1 | 4 | 388 | |||
XM_017001807.1 | XP_016857296.1 | 4 | 419 | |||
XM_017001808.1 | XP_016857297.1 | 4 | 422 | |||
GenBank mRNA | AB037846.1 | - | 4 | 328 | ||
AF265228.1 | - | 3 | 243 | |||
AK021605.1 | - | 4 | 307 | |||
AK289366.1 | - | 4 | 346 | |||
AK293598.1 | - | 4 | 354 | |||
AK300217.1 | - | 4 | 321 | |||
AK300372.1 | - | 4 | 313 | |||
AK300442.1 | - | 5 | 429 | |||
AL136657.1 | - | 4 | 325 | |||
AL137200.1 | - | 4 | 327 | |||
BC002855.2 | - | 4 | 312 | 3635724 | ||
BC008053.2 | - | 3 | 506 | 3834056 | ||
CA842195.1 | - | 4 | 355 |