Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018838.4 | NP_061326.1 | 2, 3 | 213 | ||
GenBank mRNA | AF112208.1 | - | 2, 3 | 233 | ||
AF217092.1 | - | 2, 3 | 192 | |||
AK222619.1 | - | 2, 3 | 200 | |||
BC005936.1 | - | 2, 3 | 202 | 4069306 | ||
BG943528.1 | - | 2, 3 | 198 | |||
BM548072.1 | - | 280 | ||||
BT007220.1 | - | 2, 3 | 180 | |||
U34343.1 | - | 319 |