Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022052.1 | NP_071335.1 | 13, 14 | 1240 | ||
GenBank mRNA | AF346619.1 | - | 13, 14 | 1240 | ||
AJ277527.1 | - | 13, 14 | 1240 | |||
AJ277660.1 | - | 10, 11 | 798 | |||
AK057086.1 | - | 2, 3 | 1623 | |||
AK294826.1 | - | 267 | ||||
BC031616.1 | - | 13, 14 | 1233 | 5170537 |